Learn More
2-Methylbenzyl chloride, 98+%, Thermo Scientific Chemicals
£10.25 - £166.00
Chemical Identifiers
CAS | 552-45-4 |
---|---|
Molecular Formula | C8H9Cl |
Molecular Weight (g/mol) | 140.61 |
MDL Number | MFCD00000899 |
InChI Key | VQRBXYBBGHOGFT-UHFFFAOYSA-N |
Synonym | 2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw |
PubChem CID | 11093 |
IUPAC Name | 1-(chloromethyl)-2-methylbenzene |
SMILES | CC1=CC=CC=C1CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495503
|
Thermo Scientific Alfa Aesar
A13448.14 |
25 g | |||||||
11405513
|
Thermo Scientific Alfa Aesar
A13448.22 |
100 g |
£34.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415513
|
Thermo Scientific Alfa Aesar
A13448.36 |
500 g |
£166.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
552-45-4 | |
140.61 | |
VQRBXYBBGHOGFT-UHFFFAOYSA-N | |
11093 | |
CC1=CC=CC=C1CCl |
C8H9Cl | |
MFCD00000899 | |
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw | |
1-(chloromethyl)-2-methylbenzene |
Specifications
552-45-4 | |
197°C to 199°C | |
Pungent | |
MFCD00000899 | |
386016 | |
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw | |
CC1=CC=CC=C1CCl | |
140.61 | |
140.61 | |
2-Methylbenzyl chloride |
2°C | |
73°C (163°F) | |
C8H9Cl | |
UN3265 | |
14,10091 | |
VQRBXYBBGHOGFT-UHFFFAOYSA-N | |
1-(chloromethyl)-2-methylbenzene | |
11093 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
Recommended Storage : 2° to 8°C
RUO – Research Use Only