missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl mercaptan, 97%, Thermo Scientific Chemicals

Product Code. 11422488
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7341-24-4
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00022074
InChI Key PJUDFYDAJBQPEA-UHFFFAOYSA-N
Synonym 2-methylphenyl methanethiol, 2-methylbenzyl mercaptan, 2-methylbenzylmercaptan, 2-methylbenzenemethanethiol, o-methylbenzyl mercaptan, o-tolylmethanethiol, o-tolyl-methanethiol, 2-methylphenyl methane-1-thiol
PubChem CID 288028
IUPAC Name (2-methylphenyl)methanethiol
SMILES CC1=CC=CC=C1CS

Specifications

Boiling Point 56°C to 58°C (5 mmHg)
Odor Stench
Refractive Index 1.572
Quantity 25 g
UN Number UN2810
Beilstein 2040956
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 2-Methylbenzyl mercaptan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'