missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, 99+%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10665222
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Acidity 0.0005 meq/g max.
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 240nm, 0.05 max. at 230nm, 0.07 max. at 220nm, 0.3 max. at 210nm, 1 max. at 192nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point -160.0°C
Density 0.6200g/mL
Boiling Point 30.0°C
Flash Point −51°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)2
Refractive Index 1.3530 to 1.3550
Quantity 25 mL
Beilstein 01, 134
Residue after Evaporation 0.0003% max.
Specific Gravity 0.62
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol
Formula Weight 72.15
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 2-Methylbutane, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylbutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H224-Extremely flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'