missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutylamine, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | C2H5CH(CH3)CH2NH2 |
Quantity | 1 g |
Formula Weight | 87.16 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methylbutylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'