missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutylamine, 95%, Thermo Scientific™

Product Code. 10548744
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 96-15-1
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008147
InChI Key VJROPLWGFCORRM-UHFFFAOYSA-N
Synonym 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
PubChem CID 7283
IUPAC Name 2-methylbutan-1-amine
SMILES CCC(C)CN

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula C2H5CH(CH3)CH2NH2
Quantity 25 g
Formula Weight 87.16
Percent Purity 95%
Chemical Name or Material 2-Methylbutylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'