Learn More
2-Methylbutylamine, 95%, Thermo Scientific™
£9.20 - £81.70
Chemische Identifikatoren
CAS | 96-15-1 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD00008147 |
InChI Key | VJROPLWGFCORRM-UHFFFAOYSA-N |
Synonym | 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine |
PubChem CID | 7283 |
IUPAC Name | 2-methylbutan-1-amine |
SMILES | CCC(C)CN |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10125772
|
Thermo Scientific Maybridge
SB01376DA |
1 g | Amber Glass Bottle |
£9.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10452733
|
Thermo Scientific Maybridge
SB01376EA |
10 g | Amber Glass Bottle |
£42.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10548744
|
Thermo Scientific Maybridge
SB01376EB |
25 g | Amber Glass Bottle |
£81.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Chemische Identifikatoren
96-15-1 | |
87.166 | |
VJROPLWGFCORRM-UHFFFAOYSA-N | |
7283 | |
CCC(C)CN |
C5H13N | |
MFCD00008147 | |
2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine | |
2-methylbutan-1-amine |
Spezifikation
96-15-1 | |
95% | |
C2H5CH(CH3)CH2NH2 | |
2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine | |
CCC(C)CN | |
87.166 | |
87.16 | |
2-Methylbutylamine |
95.0 | |
C5H13N | |
MFCD00008147 | |
VJROPLWGFCORRM-UHFFFAOYSA-N | |
2-methylbutan-1-amine | |
7283 | |
95% |
Sicherheit und Handhabung
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger