missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

Product Code. 10551462
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-60-8
InChI Key LFSAPCRASZRSKS-UHFFFAOYSA-N
Synonym 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
PubChem CID 11419
IUPAC Name 2-methylcyclohexan-1-one
SMILES CC1CCCCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless to Yellow
Density 0.9240g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O
Linear Formula CH3C6H9(=O)
Refractive Index 1.4470 to 1.4490
Quantity 50 g
Beilstein 07, 14
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Molecular Weight (g/mol) 112.17
Formula Weight 112.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.