missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals

Product Code. 11359226
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.

Solubility
Insoluble in water. Soluble in methanol and diethyl ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-60-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00001635
InChI Key LFSAPCRASZRSKS-UHFFFAOYSA-N
Synonym 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone
PubChem CID 11419
IUPAC Name 2-methylcyclohexan-1-one
SMILES CC1CCCCC1=O

Specifications

Melting Point °C to 14°C
Density 0.925
Boiling Point 162°C to 163°C
Flash Point 46°C (114°F)
Assay Percent Range 98%
Refractive Index 1.448
Quantity 100 mL
UN Number UN2297
Beilstein 506751
Solubility Information Insoluble in water. Soluble in methanol and diethyl ether.
Formula Weight 112.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'