missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexylamine, cis + trans, 97%, Thermo Scientific Chemicals

Product Code. 11397645
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7003-32-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001492
InChI Key FEUISMYEFPANSS-UHFFFAOYNA-N
Synonym 2-methylcyclohexylamine, cyclohexanamine, 2-methyl, o-methylcyclohexylamine, cyclohexylamine, 2-methyl, 2-methylcyclohexanamine, 2-methylcyclohexylamine,c&t, hexahydro-o-toluidine, 2methylcyclohexylamine, 2-methyl-cyclohexylamine, 2-methylcyclohexyl amine
PubChem CID 23432
IUPAC Name 2-methylcyclohexan-1-amine
SMILES CC1CCCCC1N

Specifications

Density 0.858
Boiling Point 149°C to 150°C
Flash Point 21°C (69°F)
Refractive Index 1.4565
Quantity 50 g
UN Number UN2733
Beilstein 2689073
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.2
Percent Purity 97%
Chemical Name or Material 2-Methylcyclohexylamine, Cis + Trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.