missing translation for 'onlineSavingsMsg'
Learn More

2-Methylformanilide, 98%, Thermo Scientific™

Product Code. 11486187
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-69-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00014122
InChI Key ZXTLGJAARBNQGK-UHFFFAOYSA-N
Synonym 2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
PubChem CID 7202
IUPAC Name N-(2-methylphenyl)formamide
SMILES CC1=CC=CC=C1NC=O

Specifications

Melting Point 58°C to 60°C
Boiling Point 287°C to 289°C
Quantity 100 g
Beilstein 2205748
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 2-Methylformanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'