missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11324806
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41693-47-4
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD01318644
InChI Key WKEBLSOTVYFYKY-UHFFFAOYNA-N
Synonym 2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
PubChem CID 3458543
IUPAC Name 2-methylhexanoyl chloride
SMILES CCCCC(C)C(Cl)=O

Specifications

Density 0.957
Boiling Point 163°C
Flash Point 55°C (131°F)
Refractive Index 1.429
Quantity 10 g
UN Number UN2920
Sensitivity Moisture sensitive
Formula Weight 148.63
Percent Purity 97%
Chemical Name or Material 2-Methylhexanoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'