missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoyl chloride, 97%, Thermo Scientific Chemicals

£30.70 - £196.00

Chemical Identifiers

CAS 41693-47-4
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD01318644
InChI Key WKEBLSOTVYFYKY-UHFFFAOYNA-N
Synonym 2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
PubChem CID 3458543
IUPAC Name 2-methylhexanoyl chloride
SMILES CCCCC(C)C(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334806
View Documents
Thermo Scientific Alfa Aesar
H32829.03
1 g
£30.70
1g
Please to purchase this item. Need a web account? Register with us today!
11324806
View Documents
Thermo Scientific Alfa Aesar
H32829.09
10 g
£196.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41693-47-4
148.63
WKEBLSOTVYFYKY-UHFFFAOYNA-N
3458543
CCCCC(C)C(Cl)=O
C7H13ClO
MFCD01318644
2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
2-methylhexanoyl chloride

Specifications

41693-47-4
163°C
C7H13ClO
MFCD01318644
Moisture sensitive
WKEBLSOTVYFYKY-UHFFFAOYNA-N
2-methylhexanoyl chloride
3458543
97%
0.957
55°C (131°F)
1.429
UN2920
2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
CCCCC(C)C(Cl)=O
148.63
148.63
2-Methylhexanoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only