missing translation for 'onlineSavingsMsg'
Learn More
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10147752

2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10147752
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10147752

Brand: Thermo Scientific Maybridge CC21701CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21801-79-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD00244615
InChI Key FBIUGCLQMKPURJ-UHFFFAOYSA-N
Synonym 2-methylimidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl, 2-methyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, pubchem16305, imidazo 1, 2-methyl, 2-methylimidazo 1,2-a nicotinic acid, 3-carboxy-2-methylimidazo 1,2-a pyridine
PubChem CID 89054
IUPAC Name 2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES CC1=C(N2C=CC=CC2=N1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 2-Methylimidazo[1, 2-a]pyridine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.