missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Product Code. 15467467
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

2-Methylimidazo[1,2-a]pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazo[1,2-a]pyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 934-37-2
Melting Point 39°C to 42°C
Boiling Point 148°C (20 mmHg)
Molecular Formula C8H8N2
MDL Number MFCD02251330
Quantity 1 g
Sensitivity Very Hygroscopic
Synonym 2-methylimidazo 1,2-a pyridine, 2-methyl-imidazo 1,2-a pyridine, 2-methylh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 2-methyl, methylimidazo 1,2-a pyridine, 2-methyl-4-hydroimidazo 1,2-a pyridine, imidazo 1,2-a pyridine,2-methyl
Solubility Information Slightly soluble in water.
InChI Key BZACBBRLMWHCNM-UHFFFAOYSA-N
SMILES CC1=CN2C=CC=CC2=N1
IUPAC Name 2-methylimidazo[1,2-a]pyridine
Molecular Weight (g/mol) 132.166
PubChem CID 136742
Formula Weight 132.17
Percent Purity 95%
Chemical Name or Material 2-Methylimidazo[1,2-a]pyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.