Learn More
2-Methylimidazole, 99%, Thermo Scientific Chemicals
£28.40 - £136.00
Chemical Identifiers
CAS | 693-98-1 |
---|---|
Molecular Formula | C4H6N2 |
Molecular Weight (g/mol) | 82.11 |
MDL Number | MFCD00005190 |
InChI Key | LXBGSDVWAMZHDD-UHFFFAOYSA-N |
Synonym | 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107 |
PubChem CID | 12749 |
IUPAC Name | 2-methyl-1H-imidazole |
SMILES | CC1=NC=CN1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10336530
|
Thermo Scientific Acros
127001000 |
100 g | Plastic bottle |
£28.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10346530
|
Thermo Scientific Acros
127005000 |
500 g | Plastic bottle |
£81.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061190
|
Thermo Scientific Acros
127000010 |
1 kg | Plastic bottle |
£136.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
693-98-1 | |
82.11 | |
LXBGSDVWAMZHDD-UHFFFAOYSA-N | |
12749 | |
CC1=NC=CN1 |
C4H6N2 | |
MFCD00005190 | |
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107 | |
2-methyl-1H-imidazole |
Specifications
693-98-1 | |
100.0 | |
White to Yellow | |
155°C | |
98.5% min. (GC) | |
MFCD00005190 | |
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107 | |
LXBGSDVWAMZHDD-UHFFFAOYSA-N | |
2-methyl-1H-imidazole | |
12749 | |
99% | |
2-Methylimidazole, 99% |
98.5 | |
142.0°C to 145.0°C | |
270.0°C | |
Authentic | |
C4H6N2 | |
23, 65 | |
Solubility in water: 267g/L (20°C) | |
CC1=NC=CN1 | |
82.11 | |
82.1 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Suspected of causing cancer.
May damage fertility or the unborn child.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-765-7
missing translation for 'rtecsNumber' : NI7175000
missing translation for 'tsca' : TSCA
- 2-Methylimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360-May damage fertility or the unborn child.
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only