Learn More
2-Methylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
£94.63 - £292.00
Chemical Identifiers
CAS | 660867-80-1 |
---|---|
Molecular Formula | C12H18BNO2 |
Molecular Weight (g/mol) | 219.09 |
MDL Number | MFCD08061590 |
InChI Key | MBTULFIFECUURA-UHFFFAOYSA-N |
Synonym | 2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester |
PubChem CID | 16414274 |
IUPAC Name | 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
SMILES | CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
660867-80-1 | |
219.09 | |
MBTULFIFECUURA-UHFFFAOYSA-N | |
16414274 | |
CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1 |
C12H18BNO2 | |
MFCD08061590 | |
2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester | |
2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
660867-80-1 | |
94% min. (GC) | |
C12H18BNO2 | |
2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester | |
MBTULFIFECUURA-UHFFFAOYSA-N | |
2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
16414274 | |
95% |
66.0°C to 71.0°C | |
Glass bottle | |
MFCD08061590 | |
Solubility in water: insoluble | |
CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1 | |
219.09 | |
219.09 | |
2-Methylpyridine-4-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
- 2-Methylpyridine-4-boronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only