missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 13571620
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660867-80-1
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD08061590
InChI Key MBTULFIFECUURA-UHFFFAOYSA-N
Synonym 2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester
PubChem CID 16414274
IUPAC Name 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 66.0°C to 71.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Formula Weight 219.09
Percent Purity 95%
Chemical Name or Material 2-Methylpyridine-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Methylpyridine-4-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'