missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrrolidine, 97%, Thermo Scientific Chemicals

£77.30 - £250.00

Chemical Identifiers

CAS 765-38-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00014491
InChI Key RGHPCLZJAFCTIK-UHFFFAOYSA-N
PubChem CID 13003
IUPAC Name 2-methylpyrrolidine
SMILES CC1CCCN1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398297
View Documents
Thermo Scientific Alfa Aesar
031902.03
1 g
£77.30
1g
Please to purchase this item. Need a web account? Register with us today!
11308307
View Documents
Thermo Scientific Alfa Aesar
031902.06
5 g
£250.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Methylpyrrolidine can be used to produce 2-(2-methylpyrrolidinyl)benzaldehyde at the temperature of 152°C. It is commonly used as organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

765-38-8
85.15
RGHPCLZJAFCTIK-UHFFFAOYSA-N
2-methylpyrrolidine
C5H11N
MFCD00014491
13003
CC1CCCN1

Specifications

765-38-8
2°C (37°F)
C5H11N
MFCD00014491
pyrrolidine, 2-methyl, 2-methyl-pyrrolidine, 2-methylpyrolidine, 2-methyl-pyrolidine, 2-methyl pyrrolidine, pubchem9410, pyrrolidine,2-methyl, acmc-209mch, acmc-1aspi, rac-2-methyl-pyrrolidine
RGHPCLZJAFCTIK-UHFFFAOYSA-N
2-methylpyrrolidine
13003
97%
2-Methylpyrrolidine
0.834 g/mL
Amine-like
1.437
UN2733
Soluble in water.
CC1CCCN1
85.15
85.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 212-144-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only