missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrrolidine hydrochloride, 97%, Thermo Scientific™

Product Code. 15532333 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15532333

Brand: Thermo Scientific Acros 458070050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54677-53-1
Molecular Formula C5H11N·HCl
Molecular Weight (g/mol) 121.61
InChI Key JNEIFWYJFOEKIM-UHFFFAOYSA-N
Synonym 2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
PubChem CID 21907341
IUPAC Name 2-methylpyrrolidine;hydrochloride
SMILES CC1CCCN1.Cl

Specifications

Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 121.61
Percent Purity 97%
Chemical Name or Material 2-Methylpyrrolidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.