missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylresorcinol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.
Solubility
Soluble in water and methanol.
Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents
2-Methylresorcinol is used in the preparation of aromatic benziporphyrins, tripyrrane analogs and C-5-bromo-2-hydroxyphenylcalix[4]-2-methylresorcinarene. Further, it is employed in oxidative and non-oxidative hair dye formulations.
Solubility
Soluble in water and methanol.
Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 118°C to 122°C |
Boiling Point | 264°C |
Flash Point | 135°C (275°F) |
Quantity | 50 g |
UN Number | UN2811 |
Beilstein | 2042177 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 124.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methylresorcinol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'