missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfinyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15449967
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-97-4
Molecular Formula C7H9BO3S
Molecular Weight (g/mol) 184.02
MDL Number MFCD03095261
InChI Key PHORKVSBWZGTEX-UHFFFAOYNA-N
Synonym 2-methylsulfinyl phenylboronic acid, 2-methylsulfinyl phenyl boronic acid, 2-methylsulfinylphenyl boronic acid, 2-methylsulphinyl benzeneboronic acid, 2-methanesulfinylphenylboronic acid, 1-borono-2-methylsulphinyl benzene, 2-methylsulfinyl benzeneboronic acid, pubchem1874, acmc-209q0z
PubChem CID 2773529
IUPAC Name (2-methylsulfinylphenyl)boronic acid
SMILES CS(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 118°C to 122°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 184.02
Percent Purity 97%
Chemical Name or Material 2-(Methylsulfinyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'