missing translation for 'onlineSavingsMsg'
Learn More

2'-(Methylsulfonyl)acetanilide, 96%, Thermo Scientific™

Product Code. 11481460
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20628-27-7
Molecular Formula C9H11NO3S
Molecular Weight (g/mol) 213.251
MDL Number MFCD01464107
InChI Key NGWCPCSZLLJKPD-UHFFFAOYSA-N
Synonym n-2-methylsulfonyl phenyl acetamide, n-2-methanesulfonylphenyl acetamide, n-2-methanesulfonyl phenyl acetamide, acmc-1ceiy, 2'-methylsulfonylacetanilide, n-2-methylsulfonylphenyl acetamide, n-acetyl-2-methylsulfonyl aniline, n-acetyl-2-methylsulphonyl aniline, 2'-methylsulfonyl acetanilide
PubChem CID 826278
IUPAC Name N-(2-methylsulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1S(=O)(=O)C

Specifications

Melting Point 146°C to 148°C
Quantity 1 g
Formula Weight 213.26
Percent Purity 96%
Chemical Name or Material 2'-(Methylsulfonyl)acetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'