missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)-4,5-dihydro-1H-imidazole hydrobromide, Thermo Scientific™

Product Code. 10523032
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 40241-78-9
Molecular Formula C4H9BrN2S
Molecular Weight (g/mol) 197.094
MDL Number MFCD01570110
InChI Key UPISCLUJZRQPPJ-UHFFFAOYSA-N
Synonym 2-methylthio-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-2-methylthio-1h-imidazole monohydrobromide, 2-methylsulfanyl-4,5-dihydro-1h-imidazole hydrobromide, 2-methylthio-2-imidazoline, bromide, 1h-imidazole,4,5-dihydro-2-methylthio-, hydrobromide 1:1
PubChem CID 2777335
IUPAC Name 2-methylsulfanyl-4,5-dihydro-1H-imidazole;hydrobromide
SMILES CSC1=NCCN1.Br

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.1
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'