missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Not miscible in water.
Notes
Do not store together with acids. Store away from Oxidizing agents.
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Not miscible in water.
Notes
Do not store together with acids. Store away from Oxidizing agents.
Specifications
Specifications
Density | 1.06 |
Boiling Point | 169°C to 171°C |
Flash Point | 70°C (158°F) |
Odor | Stench |
Linear Formula | CH3S(CH2)2OH |
Refractive Index | 1.4944 |
Quantity | 100 g |
UN Number | UN3334 |
Beilstein | 1731081 |
Solubility Information | Not miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'