missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ 2'-MOE-G Phosphoramidite, 28-400 finish bottle

2'-MOE-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis.

Brand:  Thermo Scientific™ 27-1022-82

27.62 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 17987182

  • £34.10 / 2g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

2'-MOE-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 28-400 finish neck style, which is compatible with most MerMade synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-N2-isobutyryl-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C47H60N7O10P
Base: G
Base Protection: iBu
Formula weight: 914.01 g/mol
CAS number: 251647-55-9
HPLC purity: ≥99%; M-11 Impurity ≤0.01%
31P NMR purity: ≥99%

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MerMade Synthesizers
Store in freezer (at or below -20°C).
2in-MOE modified
99%
Phosphoramidite
RNA
Powder
2in-MOE-G CEP
Guanosine (G)
Room Temperature
2 g
TheraPure™
TheraPure™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers