missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-3541145
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10154104

Brand: Thermo Scientific Maybridge CC62801CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906353-04-6
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.24
MDL Number MFCD09702410
InChI Key JXQGQYNBVNKCAK-UHFFFAOYSA-N
PubChem CID 24229698
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1=CSC(=N1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid, 2-morpholino-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-morpholinyl, 2-morpholinothiazole-4-carboxylic acid, 4-4-carboxy-1,3-thiazol-2-yl morpholine, 4-thiazolecarboxylicacid,2-4-morpholinyl, 4-carboxy-2-morpholin-4-yl-1,3-thiazole, 4-thiazolecarboxylic acid, 2-4-morpholinyl, 2-morpholin-4-yl-1,3-thiazole-4-carboxylic acid
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 2-Morpholino-1, 3-thiazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.