Learn More
(2-Morpholino-3-pyridinyl)methanol, 97%, Thermo Scientific™
£33.40 - £93.20
Chemical Identifiers
CAS | 423768-55-2 |
---|---|
Molecular Formula | C10H14N2O2 |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD03086196 |
InChI Key | WESHZTWFDUKWGL-UHFFFAOYSA-N |
PubChem CID | 2776571 |
IUPAC Name | (2-morpholin-4-ylpyridin-3-yl)methanol |
SMILES | OCC1=C(N=CC=C1)N1CCOCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10029281
|
Thermo Scientific Maybridge
CC24609CB |
250 mg | Amber Glass Bottle |
£33.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10581492
|
Thermo Scientific Maybridge
CC24609DA |
1 g | Amber Glass Bottle |
£93.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
423768-55-2 | |
194.23 | |
WESHZTWFDUKWGL-UHFFFAOYSA-N | |
(2-morpholin-4-ylpyridin-3-yl)methanol |
C10H14N2O2 | |
MFCD03086196 | |
2776571 | |
OCC1=C(N=CC=C1)N1CCOCC1 |
Specifications
423768-55-2 | |
100.0 | |
C10H14N2O2 | |
2-morpholino-3-pyridinyl methanol, 2-morpholin-4-yl pyridin-3-yl methanol, 3-pyridinemethanol,2-4-morpholinyl, 2-morpholinopyridin-3-yl methanol, 3-pyridinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyridin-3-yl methanol, 2-morpholin-4-yl-3-pyridyl methan-1-ol | |
OCC1=C(N=CC=C1)N1CCOCC1 | |
194.23 | |
194.23 | |
(2-Morpholino-3-pyridinyl)methanol |
96.5 | |
97% | |
MFCD03086196 | |
WESHZTWFDUKWGL-UHFFFAOYSA-N | |
(2-morpholin-4-ylpyridin-3-yl)methanol | |
2776571 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.