missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10782634
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 865471-20-1
Molecular Formula C12H12F3NO3
Molecular Weight (g/mol) 275.227
MDL Number MFCD09025883
InChI Key PRECFTDWCNEEDB-UHFFFAOYSA-N
Synonym 2-morpholino-5-trifluoromethyl benzoic acid, 2-morpholin-4-yl-5-trifluoromethyl benzoic acid, benzoic acid,2-4-morpholinyl-5-trifluoromethyl
PubChem CID 24229572
IUPAC Name 2-morpholin-4-yl-5-(trifluoromethyl)benzoic acid
SMILES C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 275.22
Percent Purity 97%
Chemical Name or Material 2-Morpholino-5-(trifluoromethyl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.