missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinophenol, 97%, Thermo Scientific™

Product Code. 10213472
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10213472

Brand: Thermo Scientific Maybridge S08696DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 41536-44-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00051674
InChI Key VMPYFWTYGZZUMY-UHFFFAOYSA-N
PubChem CID 2795394
IUPAC Name 2-morpholin-4-ylphenol
SMILES C1COCCN1C2=CC=CC=C2O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-morpholinophenol, 2-morpholin-4-yl phenol, 2-4-morpholino phenol, phenol, 2-4-morpholinyl, 2-4-morpholinyl phenol, 2-morpholin-4-yl-phenol, 2-4-morpholinyl-phenol, phenol,2-4-morpholinyl
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinophenol
compliance-icons
Product Identifier
  • 2-Morpholinophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.