missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Product Code. 10563032
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-33-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD03086181
InChI Key MYGVYNRBQSAMIF-UHFFFAOYSA-N
Synonym 2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
PubChem CID 2776562
SMILES OCC1=CC=CC=C1N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (2-morpholin-4-ylphenyl)methanol
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (2-Morpholinophenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'