Learn More
2-(N,N-Dimethylaminomethyl)phenylboronic acid, 98%, Thermo Scientific Chemicals
£79.20 - £361.90
Chemical Identifiers
CAS | 85107-53-5 |
---|---|
Molecular Formula | C9H14BNO2 |
Molecular Weight (g/mol) | 179.02 |
MDL Number | MFCD01318999 |
InChI Key | VUDCTLOJEPCNRS-UHFFFAOYSA-N |
Synonym | 2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid |
PubChem CID | 2734345 |
IUPAC Name | [2-[(dimethylamino)methyl]phenyl]boronic acid |
SMILES | B(C1=CC=CC=C1CN(C)C)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10251582
|
Thermo Scientific Acros
359241000 |
100 mg | Glass bottle |
£79.20
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696344
|
Thermo Scientific Acros
359240010 |
1 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85107-53-5 | |
179.02 | |
VUDCTLOJEPCNRS-UHFFFAOYSA-N | |
2734345 | |
B(C1=CC=CC=C1CN(C)C)(O)O |
C9H14BNO2 | |
MFCD01318999 | |
2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid | |
[2-[(dimethylamino)methyl]phenyl]boronic acid |
Specifications
85107-53-5 | |
100.0 | |
Authentic | |
C9H14BNO2 | |
2-dimethylamino methyl phenyl boronic acid, 2-n,n-dimethylaminomethyl phenylboronic acid, 2-dimethylamino methyl phenylboronic acid, boronic acid, 2-dimethylamino methyl phenyl, 2-n,n-dimethylaminomethyl benzeneboronic acid, 2-n,n-dimethylaminomethyl phenyl-boronic acid, 2-dimethylamino methyl phenyl boranediol, 2-dimethylamino methyl benzeneboronic acid | |
B(C1=CC=CC=C1CN(C)C)(O)O | |
179.02 | |
179.02 | |
Powder |
97.5 | |
Beige | |
97.5% min. (HPLC) | |
MFCD01318999 | |
VUDCTLOJEPCNRS-UHFFFAOYSA-N | |
[2-[(dimethylamino)methyl]phenyl]boronic acid | |
2734345 | |
98% | |
2-(N, N-Dimethylaminomethyl)phenylboronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
- 2-(N,N-Dimethylaminomethyl)phenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only