missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals
£25.50 - £340.00
Chemical Identifiers
CAS | 32316-92-0 |
---|---|
Molecular Formula | C10H9BO2 |
Molecular Weight (g/mol) | 171.99 |
MDL Number | MFCD00236051 |
InChI Key | KPTRDYONBVUWPD-UHFFFAOYSA-N |
Synonym | 2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol |
PubChem CID | 2734375 |
SMILES | OB(O)C1=CC2=CC=CC=C2C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491167
|
Thermo Scientific Alfa Aesar
B24157.03 |
1 g |
£25.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401177
|
Thermo Scientific Alfa Aesar
B24157.06 |
5 g |
£71.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411177
|
Thermo Scientific Alfa Aesar
B24157.14 |
25 g |
£340.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32316-92-0 | |
171.99 | |
KPTRDYONBVUWPD-UHFFFAOYSA-N | |
2734375 |
C10H9BO2 | |
MFCD00236051 | |
2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol | |
OB(O)C1=CC2=CC=CC=C2C=C1 |
Specifications
32316-92-0 | |
C10H9BO2 | |
2936449 | |
KPTRDYONBVUWPD-UHFFFAOYSA-N | |
naphthalen-2-ylboronic acid | |
2734375 | |
97% |
∼267°C | |
MFCD00236051 | |
2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol | |
OB(O)C1=CC2=CC=CC=C2C=C1 | |
171.99 | |
171.99 | |
2-Naphthaleneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only