missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthalenemethanol, 98%, Thermo Scientific Chemicals
£20.50 - £220.10
Chemical Identifiers
CAS | 1592-38-7 |
---|---|
Molecular Formula | C11H10O |
Molecular Weight (g/mol) | 158.2 |
MDL Number | MFCD00004124 |
InChI Key | MFGWMAAZYZSWMY-UHFFFAOYSA-N |
Synonym | 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na |
PubChem CID | 74128 |
ChEBI | CHEBI:27615 |
IUPAC Name | naphthalen-2-ylmethanol |
SMILES | C1=CC=C2C=C(C=CC2=C1)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10724743
|
Thermo Scientific Acros
351150010 |
1 g | Glass bottle |
N/A
|
N/A | ||||
10282842
|
Thermo Scientific Acros
351150050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10646564
|
Thermo Scientific Acros
351150250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1592-38-7 | |
158.2 | |
MFGWMAAZYZSWMY-UHFFFAOYSA-N | |
74128 | |
naphthalen-2-ylmethanol |
C11H10O | |
MFCD00004124 | |
2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na | |
CHEBI:27615 | |
C1=CC=C2C=C(C=CC2=C1)CO |
Specifications
1592-38-7 | |
100.0 | |
White | |
97.5% min. (GC) | |
C10H7CH2OH | |
2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na | |
C1=CC=C2C=C(C=CC2=C1)CO | |
158.2 | |
CHEBI:27615 | |
98% | |
2-Naphthalenemethanol |
97.5 | |
79°C to 83°C | |
Authentic | |
C11H10O | |
MFCD00004124 | |
MFGWMAAZYZSWMY-UHFFFAOYSA-N | |
naphthalen-2-ylmethanol | |
74128 | |
158.2 | |
Crystalline Powder |
Safety and Handling
Product Identifier
- 2-Naphthalenemethanol
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Naphthalenemethanol, 98%, Thermo Scientific Chemicals