missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11408396
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-23-0
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004066
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
PubChem CID 8422
ChEBI CHEBI:50397
IUPAC Name 2-naphthalen-2-yloxyacetic acid
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O

Specifications

Melting Point 155°C to 157°C
Quantity 2500 g
Beilstein 1074148
Merck Index 14,6397
Solubility Information Soluble in alcohol,ether,acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Naphthoxyacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'