Learn More
2-Naphthoyl chloride, 98%, Thermo Scientific Chemicals
£40.20 - £548.00
Chemical Identifiers
CAS | 2243-83-6 |
---|---|
Molecular Formula | C11H7ClO |
Molecular Weight (g/mol) | 190.626 |
MDL Number | MFCD00004093 |
InChI Key | XNLBCXGRQWUJLU-UHFFFAOYSA-N |
Synonym | 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride |
PubChem CID | 75246 |
IUPAC Name | naphthalene-2-carbonyl chloride |
SMILES | C1=CC=C2C=C(C=CC2=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457946
|
Thermo Scientific Alfa Aesar
A14046.09 |
10 g |
£40.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467946
|
Thermo Scientific Alfa Aesar
A14046.18 |
50 g |
£132.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477946
|
Thermo Scientific Alfa Aesar
A14046.30 |
250 g |
£548.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2243-83-6 | |
190.626 | |
XNLBCXGRQWUJLU-UHFFFAOYSA-N | |
75246 | |
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl |
C11H7ClO | |
MFCD00004093 | |
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride | |
naphthalene-2-carbonyl chloride |
Specifications
2243-83-6 | |
159°C to 160°C (9 mmHg) | |
Pungent | |
MFCD00004093 | |
907776 | |
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride | |
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl | |
190.626 | |
190.63 | |
2-Naphthoyl chloride |
48°C to 52°C | |
>110°C (230°F) | |
C11H7ClO | |
UN3261 | |
Moisture sensitive | |
XNLBCXGRQWUJLU-UHFFFAOYSA-N | |
naphthalene-2-carbonyl chloride | |
75246 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 218-822-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only