missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthylmagnesium bromide, 0.25M in MeTHF, Thermo Scientific™
Click to view available options
Quantity:
50 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 50 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Formula Weight | 231.37 |
Concentration or Composition (by Analyte or Components) | 0.25M in MeTHF |
Chemical Name or Material | 2-Naphthylmagnesium bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'