missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 21473-01-8, 109-99-9 |
CAS Min % | 9.76, 85.36 |
CAS Max % | 14.64, 90.24 |
Color | Brown |
Density | 0.9480g/mL |
Flash Point | −17°C |
Packaging | AcroSeal™ Glass Bottle |
Molecular Formula | C10H7BrMg |
MDL Number | MFCD01319910 |
Quantity | 50 mL |
Show More |
Safety and Handling
Product Identifier
- 2-Naphthylmagnesium bromide, 0.5M solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'