missing translation for 'onlineSavingsMsg'
Learn More
2-Nitroaniline, 98%, Thermo Scientific Chemicals Product Code.: 10143610

2-Nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 10143610
5 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10143610

Brand: Thermo Scientific Acros 128350050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-74-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007687
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
Synonym o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzeneShow MoreShow Less
PubChem CID 6946
IUPAC Name 2-nitroaniline
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Color Orange to Brown
Boiling Point 284.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 5 g
Beilstein 12,687
Merck Index 15,6669
Solubility Information Solubility in water: 1.1g/L in water (20°C). Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 138.13
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material 2-Nitroaniline, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0404026 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number128351000,128350051,128350050,128355000
Lot NumberA0429855 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number128351000,128350051,128350050,128355000
Lot NumberA0392834 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number128351000,128350051,128350050,128355000
Lot NumberA0346892 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number128351000,128350051,128350050,128355000
Lot NumberA0439130 Certificate TypeCertificate of Analysis Date18/02/2022 Catalog Number128351000,128350051,128350050,128355000
compliance-icons compliance-icons
Product Identifier
  • 2-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.