missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzonitrile, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
ET catalysis of Au25 in intramolecular cascade reaction of 2-nitrobenzonitrile, from which 2-amniobenzamide (a precursor of significant pharmaceuticals20,21) is produced with high yield. fluorodenitration of 2-nitrobenzonitrile with Rb18F in DMSO gave an 85% yield of the 18F-labelled benzonitrile after only 20 minutes at 150°C.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 107°C to 111°C |
Boiling Point | 165°C (16 mmHg) |
Odor | Almond-like |
Quantity | 5 g |
UN Number | UN3439 |
Beilstein | 638987 |
Solubility Information | Insoluble in water. |
Formula Weight | 148.12 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Nitrobenzonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'