missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrocinnamaldehyde, predominantly trans, 98%, Thermo Scientific Chemicals

Product Code. 11359569
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a actinometer for the UV-A range of photostability testing of pharmaceuticals. 2-Nitrocinnamaldehyde can be oxidized to 2-nitrocinnamic acid which can be used in the Baeyer-Emmerling indole synthesis to produce indole and substituted indoles.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1466-88-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007188
InChI Key VMSMELHEXDVEDE-HWKANZROSA-N
Synonym 2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
PubChem CID 5367122
IUPAC Name (E)-3-(2-nitrophenyl)prop-2-enal
SMILES C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]

Specifications

Melting Point 125°C to 129°C
Odor Characteristic
Quantity 25 g
Beilstein 908376
Sensitivity Air Sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 2-Nitrocinnamaldehyde, predominantly trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.