missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nonanone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
1000 g
Description
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones. Also used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples and to study binding of β-lactoglobulin to alkanones.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -15°C |
Density | 0.826 |
Boiling Point | 195°C |
Flash Point | 64°C (147°F) |
Refractive Index | 1.421 |
Quantity | 50 g |
Beilstein | 1743645 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 142.24 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'