missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanone, 99%, Thermo Scientific Chemicals

Product Code. 10123340
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-55-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009553
InChI Key VKCYHJWLYTUGCC-UHFFFAOYSA-N
Synonym 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
PubChem CID 13187
ChEBI CHEBI:77927
IUPAC Name nonan-2-one
SMILES CCCCCCCC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -21.0°C
Color Yellow
Density 0.8260g/mL
Boiling Point 188.0°C to 190.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)6COCH3
Refractive Index 1.4200 to 1.4220
Quantity 5 mL
Beilstein 01, 709
Specific Gravity 0.826
Solubility Information Solubility in water: 0.37g/L (25°C). Other solubilities: soluble in acetone
Formula Weight 142.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Nonanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'