missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Octanol, 97%, Honeywell™
0.97
£15.86 - £41.47
Chemical Identifiers
CAS | 123-96-6 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.231 |
MDL Number | MFCD00004591 |
InChI Key | SJWFXCIHNDVPSH-UHFFFAOYSA-N |
Synonym | 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol |
PubChem CID | 20083 |
ChEBI | CHEBI:37869 |
IUPAC Name | octan-2-ol |
SMILES | CCCCCCC(C)O |
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
123-96-6 | |
130.231 | |
SJWFXCIHNDVPSH-UHFFFAOYSA-N | |
20083 | |
octan-2-ol |
C8H18O | |
MFCD00004591 | |
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol | |
CHEBI:37869 | |
CCCCCCC(C)O |
Specifications
123-96-6 | |
0.819g/mL at 25°C | |
71°C (159.8°F) | |
C8H18O | |
MFCD00004591 | |
0.986g/L at 20°C in water | |
CCCCCCC(C)O | |
130.231 | |
CHEBI:37869 | |
0.97 | |
2-Octanol |
Colorless | |
174°C to 181°C | |
Unpleasant | |
CH3(CH2)5CH(OH)CH3 | |
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol | |
SJWFXCIHNDVPSH-UHFFFAOYSA-N | |
octan-2-ol | |
20083 | |
130.23g/mol | |
Liquid |
Safety and Handling
P280-P305 + P351 + P338 + P310
missing translation for 'rtecsNumber' : 204-667-0