missing translation for 'onlineSavingsMsg'
Learn More

2-Octene, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11316686
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-67-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCCC=CC

Specifications

Density 0.715
Boiling Point 123°C to 125°C
Flash Point 14°C (57°F)
Refractive Index 1.415
Quantity 5 g
UN Number UN3295
Beilstein 1719495
Formula Weight 112.22
Percent Purity 98%
Chemical Name or Material 2-Octene, Cis + Trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'