missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
500 mg
Description
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.
Notes
Hygroscopic. Store at room temperature.
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.
Notes
Hygroscopic. Store at room temperature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 208°C |
Color | White to Gray |
Density | 1.048 g/mL |
Quantity | 500 mg |
Formula Weight | 172.2 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2-Oxa-7-azaspiro[3.5]nonane hemioxalate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'