missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxetanemethanol, 96%
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.080 g/mL |
| Boiling Point | 98°C (15 mmHg) |
| Refractive Index | 1.446 |
| Quantity | 1 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 88.11 |
| Percent Purity | 96% |
| Physical Form | Liquid |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction