missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentanone ≥90%, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
- ≥95% for general laboratory use.
Specifications
CAS | 107-87-9 |
Melting Point | -78°C |
Color | Colorless |
Density | 0.809g/mL at 25°C |
Boiling Point | 101°C to 105°C |
Packaging | Glass bottle |
Molecular Formula | C5H10O |
Linear Formula | CH3COCH2CH2CH3 |
MDL Number | MFCD00009400 |
Quantity | 1 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'