missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxybenzaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Phenoxybenzaldehyde is used as a reagent in the synthesis of chiral 1,3,4-oxadiazol-2-ones as potent and highly selective FAAH inhibitors. Also used as a reagent in the synthesis of [4-(phenoxy)pyridin-3-yl]methylamines; a new class of selective noradrenaline reuptake inhibitors (NRI).
Solubility
Very slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
2-Phenoxybenzaldehyde is used as a reagent in the synthesis of chiral 1,3,4-oxadiazol-2-ones as potent and highly selective FAAH inhibitors. Also used as a reagent in the synthesis of [4-(phenoxy)pyridin-3-yl]methylamines; a new class of selective noradrenaline reuptake inhibitors (NRI).
Solubility
Very slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Very slightly soluble in water. |
Formula Weight | 198.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-Phenoxybenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'