missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 15466127
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13807-84-6
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00017297
InChI Key VMZBMTWFHYYOIN-UHFFFAOYSA-N
Synonym 2-phenoxyphenyl methanol, 2-phenoxybenzyl alcohol, o-phenoxybenzyl alcohol, phenoxyphenylmethanol, 2-phenoxybenzylalcohol, 2-phenoxybenzenemethanol, o-phenoxy benzyl alcohol, 2-phenoxy-benzenemethanol, benzenemethanol, phenoxy, 2-phenoxy-phenyl-methanol
PubChem CID 3660111
IUPAC Name (2-phenoxyphenyl)methanol
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CO

Specifications

Quantity 250 mg
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 2-Phenoxybenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'