missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanimidamide hydrochloride dihydrate, 97%, Thermo Scientific™

Product Code. 10464153
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10464153

Brand: Thermo Scientific Maybridge MO07572DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67386-38-3
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.64
MDL Number MFCD00173806
InChI Key ZLZLLGGMCNVVAS-UHFFFAOYSA-N
Synonym 2-phenoxyacetamidine hydrochloride, 2-phenoxyethanimidamide hydrochloride, 2-phenoxyacetimidamide hydrochloride, 2-phenoxy-acetamidine hcl, ethanimidamide, 2-phenoxy-, monohydrochloride, 2-phenoxyethanamidine, chloride, 2-phenoxyacetamidinehydrochloride, 2-phenoxy acetamidine hydrochloride, ethanimidamide,2-phenoxy-,hydrochloride 1:1
PubChem CID 2745912
IUPAC Name 2-phenoxyethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)COC1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.68
Percent Purity 97%
Chemical Name or Material 2-Phenoxyethanimidamide hydrochloride dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.